Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Hydrogen Peroxide, 6%
SCI_ED

Catalog No. S25361 Shop All Fisherbrand Products
Change view
Click to view available options
Concentration:
6% Solution
6% Solution, Stabilized
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Concentration
S25361 6% Solution, Stabilized
S25361A 6% Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25361
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Used for education and lab testing.

  • Lab Grade Solution
  • Product is for educational use
  • Available in 500 mL Container

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7722-81-1 , 7732-18-5
Synonym Hydrogen Dioxide, Hyperoxide

Specifications

Color Colorless
Linear Formula H2O2
Concentration 6% Solution, Stabilized
Density 1g/mL
Grade Laboratory
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Hydrogen Peroxide
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.