Learn More
Description
- Lab Grade Solution
- Product is for educational use
- Available in 500 mL Container
This product is for educational use - no C of A is available.
Specifications
Specifications
| Color | Colorless |
| Linear Formula | H2O2 |
| Concentration | 6% Solution |
| Density | 1g/mL |
| Grade | Laboratory |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Chemical Name or Material | Hydrogen Peroxide |
Safety and Handling
| ShelfLife | 1 year |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
