Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Hydrogen Peroxide, 6%
SCI_ED

Used for education and lab testing.

$16.67 - $27.45

Chemical Identifiers

CAS 7722-81-1 , 7732-18-5
Synonym Hydrogen Dioxide, Hyperoxide
View More Specs

Products 2
Catalog Number Concentration Price Quantity & Availability  
Catalog Number Concentration Price Quantity & Availability  
S25361A
SCI_ED
SDS View Documents
6% Solution
Each for $16.67
Edge
Add to Cart
 
 Restricted Item
S25361
SCI_ED
SDS View Documents
6% Solution, Stabilized
Each for $27.45
Case of 20 EA for $481.00
Edge
Add to Cart
 
Description

Description

  • Lab Grade Solution
  • Product is for educational use
  • Available in 500 mL Container

This product is for educational use - no C of A is available.

Specifications

Chemical Identifiers

7722-81-1, 7732-18-5
Hydrogen Dioxide, Hyperoxide

Specifications

Colorless
H2O2
Hydrogen Dioxide, Hyperoxide
500 mL
Hydrogen Peroxide
7722-81-1 , 7732-18-5
1g/mL
Laboratory
Liquid
Videos
Safety and Handling

Safety and Handling

ShelfLife : 1 year

SDS
Documents

Documents

Product Certifications
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.