Learn More
LAB ALLEY Hydrogen Peroxide 6% Solution, (Stabilized), Lab Grade 1 Liter.Recommended For Educational Or Research Laboratories.
SDP
Catalog No. NC3761967
Encompass
Hydrogen Peroxide 6% Solution, (Stabilized), Lab Grade 1 Liter.Recommended For Educational Or Research Laboratories. Hydrogen Peroxide 6% (Stabilized) is composed of 6% hydrogen peroxide (H2O2) and 94% water (H2O) with trace amount of acetic acid. Lab Grade Hydrogen Peroxide 6% is a clear, colorless liquid that contains stabilizers. Laboratory Grade Hydrogen Peroxide 6% contains intermediate purity and is suitable for educational or research laboratories; it is not safe to use in foods, drugs, or medicines. CAS Number 7722-84-1
Catalog No. NC3761967 Supplier LAB ALLEY Supplier No. HPL61L
Hydrogen Peroxide 6% Solution, (Stabilized), Lab Grade 1 Liter.Recommended For Educational Or Research Laboratories. Hydrogen Peroxide 6% (Stabilized) is composed of 6% hydrogen peroxide (H2O2) and 94% water (H2O) with trace amount of acetic acid. Lab Grade Hydrogen Peroxide 6% is a clear, colorless liquid that contains stabilizers. Laboratory Grade Hydrogen Peroxide 6% contains intermediate purity and is suitable for educational or research laboratories; it is not safe to use in foods, drugs, or medicines. CAS Number 7722-84-1