Learn More
Hydrogen peroxide, for analysis, 35 wt.% solution in water, stabilized
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Boiling Point | 108.0°C |
| Melting Point | -33.0°C |
| Color | Yellow through Brown |
| Physical Form | Powder |
| Quantity | 500 mL |
| Packaging | Plastic bottle |
| Specific Gravity | 1.135 |
| Density | 1.1350g/mL |
| Linear Formula | H2O2 |
| Fieser | 01,457; 02,216; 03,154; 04,253; 05,337; 06,286; 07,174; 08,247; 09,241; 10,201; 11,256; 12,242; 14,176; 15,166; 16,177; 17,145 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May intensify fire; oxidizer. Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye damage. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Immediately call a POISON CENTER or doctor/physician. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immedia |
| EINECSNumber | 231-765- |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.