Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, 15% (w/w), Ricca Chemical
SDP

Catalog No. R38216001B Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R38216001B 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R38216001B Supplier Ricca Chemical Company Supplier No. R38216001BV
Add to Cart
Add to Cart

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.01
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name peroxol
SMILES OO

Specifications

Color Clear colorless liquid
Assay Percent Range 15% (w/w)
Solubility Information Solubility in Water: Infinite
Percent Purity 15%
Grade Laboratory
Odor Odorless
Packaging Vented Opaque Poly Bottle
Quantity 1 L
Physical Form Solution
Chemical Name or Material Hydrogen Peroxide
Recommended Storage Oxidizer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.