Learn More
Abcam Hydrogen Pero x ide Blocking Reagent, 60ML
Catalog No. 502653805
Encompass_Preferred
Hydrogen peroxide blocking reagent is used to block endogenous peroxidase activity in IHC applications, enhancing signal detection in HRP / peroxidase-based systems. - Cited in over 30 publications - Endogenous peroxidase block – prevents non-specific background staining in IHC - HRP-based detection – ideal for use with HRP/peroxidase detection systems - Concentration – contains 0.3% hydrogen peroxide
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-265-3805 Supplier Abcam Supplier No. AB6421860ML
Hydrogen peroxide blocking reagent is used to block endogenous peroxidase activity in IHC applications, enhancing signal detection in HRP / peroxidase-based systems. - Cited in over 30 publications - Endogenous peroxidase block – prevents non-specific background staining in IHC - HRP-based detection – ideal for use with HRP/peroxidase detection systems - Concentration – contains 0.3% hydrogen peroxide
The product is subject to the following: Abcam Restricted Use Statement