Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Hydrogen Peroxide, Calbiochem™,

Catalog No. 386790500ML Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
38-679-0500ML 500 mL
38-679-0100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 38-679-0500ML Supplier MilliporeSigma™ Supplier No. 386790500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7722-84-1 | H2O2 | 34.014 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Boiling Point 141.16°C
Melting Point -11.06°C
Physical Form Liquid
Quantity 500 mL
Solubility Information Easily soluble in cold water.
Formula Weight 34
Percent Purity 29-32%
Chemical Name or Material Hydrogen Peroxide
Hazard Category H302: Harmful if swallowed.
Hazard Statement H314: Causes severe skin burns and eye damage.
Precautionary Statement H335: May cause respiratory irritation.
DOTInformation DOT Class: 5.1, Packaging Group: II
RTECSNumber MX0899000
Recommended Storage 2° to 8°C (35.6° to 46.4°F); Keep container tightly closed. Keep container in a cool, well-ventilated area. Separate from acids, alkalis, reducing agents and combustibles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.