Learn More
Specifications
Specifications
| Color | Colorless |
| Physical Form | Liquid |
| CAS Max % | 98, 2 |
| Quantity | 1 L |
| Identification | Passes Test |
| Packaging | Poly Bottle |
| Decomposition Information | Oxygen |
| Density | 1g/mL |
| Linear Formula | H2O2 |
| Solubility Information | Soluble in water |
| Show More |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement Causes skin irritation. Causes serious eye damage. |
| Precautionary Statement | GHS P Statement Wear protective gloves, eye protection. Wash exposed skin thoroughly after handling. If on skin: Wash with plenty of soap and water. If skin irritation occurs: Get medical attention. Take off contaminated clothing and wash before reuse. If in eyes: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a poison center/doctor. |
| Recommended Storage | Room Temperature |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.