Learn More
Description
- In synthetic chemistry, Hydrogen Peroxide is used for epoxidation and hydroxylation transformations.
- It is also used as the raw oxidant source for generating more complicated and specific organic/inorganic peroxides.
- Hydrogen Peroxide is a Weak Acid: H2O2 concn wt% = 35, 50, 70, 90; corresponding true pH: 4.6, 4.3, 4.4, 5.1
Specifications
Specifications
| CAS | 7722-84-1 |
| Boiling Point | 302.4°F at 760 mm Hg |
| Melting Point | 31.3°F |
| Density | 1.11 at 68°F |
| Synonym | Albone 35, Albone 50, Albone 70, Albone 35CG, Albone 50CG, Albone 70CG, Hydrogen peroxide, solution 30%, Interox, Kastone, Perone 30, Perone 35, Perone 50 |
| Solubility Information | ≥100 mg/mL at 72°F |
| Molecular Weight (g/mol) | 34.014 g/mol |
| Grade | ACS |
| Refractive Index | 1.335 |
| Vapor Pressure | 1 mm Hg at 59.5°F |
| Show More |
Safety and Handling
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.