Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, MP Biomedicals™

Catalog No. p-7236182 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-194-05780 100 mL
02-194-05790 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02-194-05780 Supplier MP Biomedicals, Inc Supplier No. 0219405780
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydrogen Peroxide is used as a reagent, disinfectant, antiseptic, and propellant.

  • In synthetic chemistry, Hydrogen Peroxide is used for epoxidation and hydroxylation transformations.
  • It is also used as the raw oxidant source for generating more complicated and specific organic/inorganic peroxides.
  • Hydrogen Peroxide is a Weak Acid: H2O2 concn wt% = 35, 50, 70, 90; corresponding true pH: 4.6, 4.3, 4.4, 5.1
TRUSTED_SUSTAINABILITY

Specifications

CAS 7722-84-1
Boiling Point 302.4°F at 760 mm Hg
Melting Point 31.3°F
Density 1.11 at 68°F
Synonym Albone 35, Albone 50, Albone 70, Albone 35CG, Albone 50CG, Albone 70CG, Hydrogen peroxide, solution 30%, Interox, Kastone, Perone 30, Perone 35, Perone 50
Solubility Information ≥100 mg/mL at 72°F
Molecular Weight (g/mol) 34.014 g/mol
Grade ACS
Refractive Index 1.335
Vapor Pressure 1 mm Hg at 59.5°F
Vapor Density 1.02
pH 4.6 to 5.1
Quantity 100 mL
Physical Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.