Learn More
World Precision Instrument Hydrogen Peroxide Sensor
SDP
Catalog No. 502521167
Encompass
The hydrogen peroxide sensors developed by WPI are designed to compliment existing high sensitivity fluorescent approaches with direct quantitative measurement in biological samples in the low nM range.
Catalog No. 50-252-1167 Supplier World Precision Instrument Supplier No. ISOHPO100LXX
The hydrogen peroxide sensors developed by WPI are designed to compliment existing high sensitivity fluorescent approaches with direct quantitative measurement in biological samples in the low nM range.