Learn More
Sigma Aldrich Fine Chemicals Biosciences Hydrogen Peroxide solutin500ML
Catalog No. 502897735
Encompass
This 30% H2O2 (w/w) is a SOLUTIONon of hydrogen peroxide. Its decomposition in the presence of varying concentrations of zeolites and manganese (catalyst) has been investigated.
Catalog No. 50-289-7735 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 31642500ML
This 30% H2O2 (w/w) is a SOLUTIONon of hydrogen peroxide. Its decomposition in the presence of varying concentrations of zeolites and manganese (catalyst) has been investigated.