Learn More
Sigma Aldrich Fine Chemicals Biosciences Hydrogen peroxide solutin500ML
Catalog No. 502897082
Encompass
The product is a 50% SOLUTIONon of hydrogen peroxide in water. Hydrogen peroxide is more commonly being used as a green oxidizing agent. It is a better alternative to other oxidants because it has high active oxygen content and the by-product is water.
Catalog No. 50-289-7082 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 516813500ML
Add to Cart
Add to Cart
The product is a 50% SOLUTIONon of hydrogen peroxide in water. Hydrogen peroxide is more commonly being used as a green oxidizing agent. It is a better alternative to other oxidants because it has high active oxygen content and the by-product is water.