Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Hydrogen Peroxide Solution, 3%
SCI_ED

Catalog No. S25359A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
500 mL
3.785 L
Packaging:
Lab Grade - 3% Solution
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25359A 3.785 L Lab Grade - 3% Solution
S25359 500 mL Lab Grade - 3% Solution
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S25359A
 Restricted Item
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Used for education and lab testing.

  • Lab Grade Solution.
  • Product is for educational use.
  • Available in 500 mL Container

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
Linear Formula H2O2
Density 1g/mL
Formula Weight 34.01
Packaging Lab Grade - 3% Solution
Quantity 3.785 L
Physical Form Liquid
Chemical Name or Material Hydrogen Peroxide Solution, 3%
Safety handling Check Alert storage code Red
ShelfLife 1 year
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.