Learn More
Sigma Aldrich Fine Chemicals Biosciences Hydrogen peroxide solution, 7722-84-1, MFCD00011333, 100mL
Catalog No. 501657229
Encompass
Empirical Formula (Hill Notation) H2O2, 30 % (w/w) in H2O, Molecular Weight 34.01, contains stabilizer
Catalog No. 50-165-7229 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. H1009100ML
Empirical Formula (Hill Notation) H2O2, 30 % (w/w) in H2O, Molecular Weight 34.01, contains stabilizer