Learn More
Sigma Aldrich Fine Chemicals Biosciences Hydrogen peroxide solution
Catalog No. 502903975
Encompass
The product is a 50% SOLUTIONon of hydrogen peroxide in water. Hydrogen peroxide is more commonly being used as a green oxidizing agent. It is a better alternative to other oxidants because it has high active oxygen content and the by-product is water.
Catalog No. 50-290-3975 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 5168134L
The product is a 50% SOLUTIONon of hydrogen peroxide in water. Hydrogen peroxide is more commonly being used as a green oxidizing agent. It is a better alternative to other oxidants because it has high active oxygen content and the by-product is water.