Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide Solution, For trace analysis, ≥30%, MilliporeSigma™ Supelco™

Catalog No. 111019705 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-9705 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-9705 Supplier MilliporeSigma Supelco Supplier No. 95321100ML
Add to Cart
Add to Cart

Specifications

CAS 7722-84-1
Vapor Pressure 23.3 mmHg (30°C)
Density 1.11 g/mL (at 20°C)
Concentration ≥30%
Linear Formula H2O2
MDL Number MFCD00011333
Quantity 100 mL
Formula Weight 34.01
Percent Purity ≥30% (titration)
Grade Trace Analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.