Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide Solution, For ultratrace analysis, ≥30% (RT), MilliporeSigma™ Supelco™

Catalog No. 111018776 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8776 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-8776 Supplier MilliporeSigma Supelco Supplier No. 16911250MLF
Add to Cart
Add to Cart

Specifications

CAS 7722-84-1
Vapor Pressure 23.3 mmHg (30°C)
Density 1.11 g/mL (at 20°C)
Linear Formula H2O2
Concentration 25-35%
MDL Number MFCD00011333
Quantity 250 mL
Formula Weight 34.01
Percent Purity ≥30% (RT)
Grade Ultratrace Analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.