Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide, Stabilized, 30%, BAKER™, J.T. Baker™

Catalog No. 02003191 Shop All Avantor Products
Change view
Click to view available options
Quantity:
500 mL
Packaging:
HDPE Pressure Bottom Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-003-191 500 mL HDPE Pressure Bottom Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-003-191 Supplier Avantor Supplier No. 218901
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Physical Form Liquid
Quantity 500 mL
Formula Weight 34.01
Percent Purity 30%
Grade ACS
Packaging HDPE Pressure Bottom Poly Bottle
Specific Gravity 1L = 1.11kg
Chemical Name or Material Hydrogen Peroxide, Stabilized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.