Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Peroxide Topical Solution, USP, 2.5-3.5%, Spectrum™ Chemical
SDP

Catalog No. 18603258 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
16 oz.
1 gal.
Packaging:
Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-603-258 16 oz. Poly Bottle
18-603-260 1 gal. Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-603-258 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. HY11016OZ

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

HY110, 7722-84-1, H2O2

Spectrum™ Chemical Hydrogen Peroxide Topical Solution, USP is a mild antiseptic used on the skin to prevent infection of minor cuts, scrapes, and burns. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.01
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
IUPAC Name peroxol
SMILES OO

Specifications

CAS Max % 100%
Percent Purity 2.5 to 3.5%
Grade USP
Packaging Poly Bottle
Quantity 16 oz.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.