Learn More
CHEMetrics™ by AquaPhoenix Hydrogen Peroxide Water Quality Visual Test Kit 0.1 to 1.0% (up to 20% with dilution)
SDP
Catalog No. 13299662
Change view
Click to view available options
No. of Tests:
30
Catalog No. No. of Tests
13-299-662 30
1 options
Catalog No. 13-299-662 Supplier CHEMetrics™ by AquaPhoenix Supplier No. K5530
Add to Cart
Add to Cart
Hydrogen peroxide test kit utilizing Titrets ampoules and reverse titration technology. The titrant volume remains fixed while the sample volume is varied.
  • This Hydrogen Peroxide Titration Kit (K-5530) comes in a cardboard box and contains everything needed to perform 30 tests (except distilled water): thirty ampoules with valve assemblies, 1.0 mL syringe, 3.0 mL syringe, titrettor, 25 mL sample cup, and instructions.
Specifications
Analytes Hydrogen Peroxide
pH Range 7
Volume (Metric) 0.5 mL
No. of Tests 30
Range 0.1 to 1.0%
Type Water Quality Visual Test Kit