Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Hydrogen Sulfide Solution

Catalog No. 742546100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-254-6100ML 100 mL
74-254-625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 74-254-6100ML Supplier Sigma Aldrich Supplier No. 742546100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Hydrogen sulfide solution can be used in the preparation of lead sulfide (PbS), titanium disulfide(TiS2) and cadmium sulfide (CdS). It is also used as a ligand in the synthesis of ammineruthenium complexes.

Specifications

CAS 7783-06-4
Quantity 100 mL
Molecular Formula H2S
Linear Formula H2S
MDL Number MFCD00011444
Molecular Weight (g/mol) 34.08 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.