Learn More
Hach Company Hydrogen Sulfide Test Kit
Supplier: Hach Company 2537801
Hydrogen Sulfide Test Kit-Model HS-C1Includes reaction vial-test papers-comparator chart and instructions-in a cardboard box Effervescent tablets are supplied by the customer Resulting from the anaerobic decomposition of organic matter hydrogen sulfide is present in many water supplies Sewage and industrial wastes are other sources of sulfide pollution Highly toxic this compound has a characteristic rotten egg odor which can be detected long before harmful concentrations are reached In the HS-C kit sulfide discolors a detection paper Results are obtained by comparison of the color to a printed chart