Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen Sulfide Water, Saturated, Ricca Chemical
SDP

Catalog No. 38504 Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
120 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3850-4 120 mL
3850-32 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 3850-4 Supplier Ricca Chemical Company Supplier No. 38504
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Meets USP and ACS RS Specifications

Chemical Identifiers

CAS 7732-18-5
Synonym dihydrogen oxide, dihydrogen monoxide

Specifications

CAS Min % 98.0
CAS Max % 100.0
Grade Laboratory
Packaging Amber Glass Bottle
pH 4.5
Quantity 120 mL
Chemical Name or Material HYDROGEN SULFIDE WATER
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.