Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hydrogen tetrabromoaurate(III) hydrate, Premion™, 99.99% (metals basis), Au 32% min

Catalog No. AA4043306
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4043306 5 g
AA4043303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4043306 Supplier Thermo Scientific Chemicals Supplier No. 04043306
Add to Cart

missing translation for 'validMainframeMsg'

Au{C(NHMe) 2 } 2 ]Cl·H 2 O and [Au{C(NHMe) 2 } 2 ]Br·H 2 O were obtained by adding methyl isocyanide and 40% aqueous methylamine successively to aqueous solutions of hydrogen tetrachloroaurate(III) hydrate and hydrogen tetrabromoaurate(III) hydrate, respectively. To analyze the influence of the bromide ions in the synthesis of the gold nanorods Hydrogen tetrabromoaurate(III) hydrate is used. Hydrogen tetrabromoaurate(III) hydrate was used for purification in continuous microfluidic synthesis and functionalization of gold nanorods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number MFCD00054118
Synonym Bromoauric acid

Specifications

Melting Point ∼27°C
Physical Form Crystalline
Quantity 5 g
Sensitivity Hygroscopic
UN Number UN3260
Solubility Information Soluble in water and alcohol
Formula Weight 517.61 (Anhydrous)
Percent Purity 99.99%
Grade Premion
Concentration or Composition (by Analyte or Components) Au 32% min.
Assay (metals basis)
Chemical Name or Material Hydrogen tetrabromoaurate(III) hydrate
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.