Learn More
Strem, An Ascensus Company CAS# 27988-77-8. 5g. Hydrogen tetrachloroaurate (III) hydrate (99.8%-Au) (min. 49% Au)(Chloroauric acid). MFCD00149903
SDP
Catalog No. 5090113390
Encompass
CAS# 27988-77-8. 5g. Hydrogen tetrachloroaurate (III) hydrate (99.8%-Au) (min. 49% Au)(Chloroauric acid). MFCD00149903. Molecular Weight: 339.79. Molecular Formula: HAuCl4∙XH2O. Color/form: yellow to orange xtl. Strem# 79-0500. http://www.strem.com/catalog/v/79-0500/
Catalog No. 50-901-13390 Supplier Strem, An Ascensus Company Supplier No. 7905005G
CAS# 27988-77-8. 5g. Hydrogen tetrachloroaurate (III) hydrate (99.8%-Au) (min. 49% Au)(Chloroauric acid). MFCD00149903. Molecular Weight: 339.79. Molecular Formula: HAuCl4∙XH2O. Color/form: yellow to orange xtl. Strem# 79-0500. http://www.strem.com/catalog/v/79-0500/