Learn More
Medchemexpress LLC Hydrogenated soya phosphatidylcholine | 97281-48-6 | 99.3% | 250 MG
SDP
Catalog No. 5000403281
Encompass_Preferred
Hydrogenated soya phosphatidylcholine is a natural product. It can extend drug release concerning drug loading and solubility for oral drug delivery of water-soluble drugs. In vitro, it can achieve an 80% encapsulation rate of Oxymatrine (HY-N0158) at an agent/HSPC ratio of 0.2, above 50°C. In vivo, it can significantly prolong the half-life of Oxymatrine, improve its bioavailability, enhance liver targeting, reduce liver damage, and promote liver recovery in the Kunming mouse acute liver injury model.
  • Natural product
  • Extends drug release for oral delivery of water-soluble drugs
  • Achieves high encapsulation rates
  • Improves bioavailability and half-life of certain drugs
  • Enhances liver targeting and reduces liver damage
  • Promotes liver recovery
Catalog No. 50-004-03281 Supplier Medchemexpress LLC Supplier No. 250MGHY42990
Hydrogenated soya phosphatidylcholine is a natural product. It can extend drug release concerning drug loading and solubility for oral drug delivery of water-soluble drugs. In vitro, it can achieve an 80% encapsulation rate of Oxymatrine (HY-N0158) at an agent/HSPC ratio of 0.2, above 50°C. In vivo, it can significantly prolong the half-life of Oxymatrine, improve its bioavailability, enhance liver targeting, reduce liver damage, and promote liver recovery in the Kunming mouse acute liver injury model.
  • Natural product
  • Extends drug release for oral delivery of water-soluble drugs
  • Achieves high encapsulation rates
  • Improves bioavailability and half-life of certain drugs
  • Enhances liver targeting and reduces liver damage
  • Promotes liver recovery