Learn More
Medchemexpress LLC Hydrogenated soya phosphatidylcholine | 97281-48-6 | 99.3% | 790.16 | 100 MG
SDP
Catalog No. 5000403897
Encompass_Preferred
Hydrogenated soya phosphatidylcholine is a natural product that can extend drug release concerning drug loading and solubility for oral drug delivery of water-soluble drugs.
  • Can achieve 80% encapsulation rate of Oxymatrine (HY-N0158) in vitro.
  • Can significantly prolong the half-life of Oxymatrine, improve its bioavailability, enhance liver targeting, reduce liver damage, and promote liver recovery in vivo.
Catalog No. 50-004-03897 Supplier Medchemexpress LLC Supplier No. HY42990100MG
Hydrogenated soya phosphatidylcholine is a natural product that can extend drug release concerning drug loading and solubility for oral drug delivery of water-soluble drugs.
  • Can achieve 80% encapsulation rate of Oxymatrine (HY-N0158) in vitro.
  • Can significantly prolong the half-life of Oxymatrine, improve its bioavailability, enhance liver targeting, reduce liver damage, and promote liver recovery in vivo.