Learn More
LAB ALLEY Lab Alley Hydrogen Peroxide 6% Solution Food Grade Kosher Halal
SDP
Supplier:  LAB ALLEY HPF61LCS
Encompass
Hydrogen Peroxide 6% is composed of 6% hydrogen peroxide (H2O2) and 94% water (H2O). Food grade hydrogen peroxide 6% is a clear colorless liquid that is free of any stabilizers or additives. Certified by the Food Chemical Codex (FCC) the hydrogen peroxide is validated for its quality and purity. Through this certification customers are assured that Lab Alley FCC certified food grade hydrogen peroxide 6% maintains integrity. Lab Alley sources hydrogen peroxide through reputable manufacturers allowing the highest quality of hydrogen peroxide 6% solution to be provided to our customers. We have a consistent quality product and are the reliable choice for your Food Grade Hydrogen Peroxide 6% Solution needs. With quick processing Lab Alley is where to buy food grade hydrogen peroxide 6%. Food grade hydrogen peroxide 6% is in stock in multiple sizes ranging from 500mL bottles to 55 gallon drums. Lab Alley also provides pallets of chemicals.
Catalog No. NC3377558