Learn More
Contec™ HydroKlean Solution
BIOPHARMAGREENER_CHOICE
Catalog No. 19456889 Shop All Contec Inc. Products
Change view
Click to view available options
Volume (Metric):
0.5 L
5 L
Quantity:
2/Cs.
8/Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Quantity
19-456-889 5 L 2/Cs.
19-456-888 0.5 L 8/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-456-889 Supplier Contec™ Supplier No. SBC170HKLE
Add to Cart
Add to Cart
This item is not returnable. View return policy
Sterile fill low endotoxin cleaner with hydrogen peroxide
Features:
  • Blend of 6% hydrogen peroxide and water for injection in a 17 oz. and 34 oz. (1 L) bag in bottle with installed trigger spray and 170 oz. (5 L) capped container
  • Ready to use and generates no Volatile Organic Compounds (VOCs)
  • Hydrogen peroxide breaks down to water and oxygen in use, so it leaves little to no residue on a surface and is sutible for product contact areas
  • Features a certified endotoxin level of <0.25 EU/ml making it ideal for use in product contact areas
  • 0.2μm filtered and sterile filled into pre-irradiated containers under Grade A unidirectional airflow and bagged in a Grade A or ISO Class 5 cleanroom
  • Bag in bottle system protects contents during use
  • Certificates of analysis are available for each batch
  • 6% hydrogen peroxide is not classed as corrosive and can be used safely in all areas of a cleanroom
  • Nonflammable and ideal for use across large areas of a cleanroom or where sparks may be an issue
  • Very low residue and eliminates the need for a residue removal stage with alcohol or water
  • Double or triple bagged allowing for ease of entry into controlled environments
  • Linear tear bags are easy to open even when wearing gloves
  • Endotoxin level of less than 0.25 EU/mL
  • Sterile filled into pre-irradiated containers in an ISO Class 5 cleanroom makes it suitable for use in all areas of the cleanroom including ISO Class 5 cabinets and RABS
  • Double bagged or triple bagged in linear tear packaging facilitates transfer into isolators and RABS
  • Trigger spray prevents bottle contents from being contaminated during use
  • Not classified as corrosive can be used safely in all areas of the cleanroom
Specifications
Sterility Sterile
Quantity 2/Cs.
Volume (English) 1.3 gal.
Volume (Metric) 5 L