Learn More
Waters Corp Waters Primer, Amino Acid, Hydrolysis, Analysis
Supplier:  Waters Corp 715006455
Encompass_Preferred
This document provides guidelines for the accurate separation, identification, and quantitation of amino acids in food, feed, and biotherapeutic products. It emphasizes the importance of adequate sample hydrolysis as a crucial first step for analyzing bound amino acids in peptides and proteins.
  • Explains that hydrolysis enables the analysis of released amino acids.
  • Covers separation using ion-exchange or reversed-phase chromatographic methods.
  • Serves as a starting point, with modifications potentially necessary depending on the sample type and its complexity.
  • Comprises 69 pages.
  • Authored by Paula Hong, Donna Johnson, Donald A. Trinite, Bill Warren, Ning Zhang.
  • First edition.
  • Available in English.
Catalog No. 50-203-9912