Learn More
Cole-Parmer Baume Hydrometer, 0/50 Degree
Supplier:  Cole-Parmer 0829961
Encompass_Preferred
H-B Instrument B61804-0900 DURAC 0/50 Degree Baume Hydrometer. NIST TraceableTemperature of Standardization 60F/60FMetal BallastIndividually Serialized.For liquids heavier than water, each hydrometer is calibrated in our triple accredited ISO/IEC 17025:2005, A2LA accredited laboratory against equipment whose calibration is traceable to NIST. Individually packaged with a multi-language Statement of Accuracy indicating accuracy traceable to NIST.
Catalog No. NC0629822