Learn More
Hach Company Hydrometer, Glass
Supplier:  Hach Company 192200
Encompass
For measuring the specific gravity of heavy liquids. Approximate length, 150 mm (6"). Single scale, 1.000 to 1.225 specific gravity with 0.005 divisions. 100 mL graduated cylinder sold separately.
Catalog No. 50-199-4641