Learn More
COLE PARMER INSTRUMENT COMPANY Hydrometer 10-70 Degree
Catalog No. NC3653064
Encompass_Preferred
Cole-Parmer 0.700/1.000 Specific Gravity and 10/70 Degree Baume Dual Scale Hydrometer for Liquids Lighter Than Water
Catalog No. NC3653064 Supplier COLE PARMER INSTRUMENT COMPANY Supplier No. UX0829963
Cole-Parmer 0.700/1.000 Specific Gravity and 10/70 Degree Baume Dual Scale Hydrometer for Liquids Lighter Than Water