Learn More
Thermco Products Inc Plastic hydrometer for liquids heavier than water (Baume), 19-31, 0.2 div, 250mm
Catalog No. NC0429891
Encompass_Preferred
Made from virgin polycarbonate. Shatter proof with clarity like glass Calibrated against N.I.S.T. standards. May be certified and traceable to N.I.S.T. Approximately 250mm
Catalog No. NC0429891 Supplier Thermco Products Inc Supplier No. ACC8503PC
Made from virgin polycarbonate. Shatter proof with clarity like glass Calibrated against N.I.S.T. standards. May be certified and traceable to N.I.S.T. Approximately 250mm