Learn More
Robinson Manufacturing HYDROMETER,ASTM 52HL,API 9/21
Catalog No. NC2221950
Encompass_Preferred
Hydrometer, ASTM 52HL, API 9/21, 15" Combined Form, 0/150F
This item is not returnable. View return policy
Catalog No. NC2221950 Supplier Robinson Manufacturing Supplier No. 403A022
Hydrometer, ASTM 52HL, API 9/21, 15" Combined Form, 0/150F