Learn More
Thermco Products Inc hydrometer,ASTM 54HM 29-41 deg 0.1 deg
Catalog No. NC1362375
Encompass_Preferred
High precision hydrometer that meet or exceed the specifications of ASTM and API. The hydrometer has been calibrated against N.I.S.T. standards. This combined form hydrometer with a spirit filled thermometer in the body with a range of 0° to 150°F and is calibrated at 60°F. length is 380mm. The hydrometer is eligable for certification.
Catalog No. NC1362375 Supplier Thermco Products Inc Supplier No. GW054HMS
High precision hydrometer that meet or exceed the specifications of ASTM and API. The hydrometer has been calibrated against N.I.S.T. standards. This combined form hydrometer with a spirit filled thermometer in the body with a range of 0° to 150°F and is calibrated at 60°F. length is 380mm. The hydrometer is eligable for certification.