Learn More
Vee Gee Hydrometer Jar, 1000 mL
Catalog No. 502250363
Encompass_Preferred
The Hydrometer Jar is made of borosilicate glass, the jar features a beaded rim and a circular base. Sized properly to eliminate surface tension effects that lead to inaccurate readings.• Maximum hydrometer length: 400 mm : 1000 mL
This item is not returnable. View return policy
Catalog No. 50-225-0363 Supplier Vee Gee Supplier No. 210501000
The Hydrometer Jar is made of borosilicate glass, the jar features a beaded rim and a circular base. Sized properly to eliminate surface tension effects that lead to inaccurate readings.• Maximum hydrometer length: 400 mm : 1000 mL