Learn More
Vee Gee Hydrometer Jar, 250 mL
Catalog No. 502250365
Encompass_Preferred
The Hydrometer Jar is made of borosilicate glass, the jar features a beaded rim and a circular base. Sized properly to eliminate surface tension effects that lead to inaccurate readings.• Maximum hydrometer length: 280 mm : 250 mL
This item is not returnable. View return policy
Catalog No. 50-225-0365 Supplier Vee Gee Supplier No. 21050250
The Hydrometer Jar is made of borosilicate glass, the jar features a beaded rim and a circular base. Sized properly to eliminate surface tension effects that lead to inaccurate readings.• Maximum hydrometer length: 280 mm : 250 mL