Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ertco™ Hydrometers

Catalog No. 15059785
Change view
Click to view available options
For Use With (Application):
Liquids heavier than Water
Specific Gravity Range:
1 to 1.22
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application) Specific Gravity Range
15-059-785 Liquids heavier than Water 1 to 1.22
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. 15-059-785 Supplier Ertco™ Supplier No. 11373

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

Specifications

Increments 0.005
Length (Metric) 150 mm
For Use With (Application) Liquids heavier than Water
Specific Gravity Range 1 to 1.22
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.