Learn More
Electron Microscopy Sciences Hydromount™ Nonfluorescing Mounting Media 100 ML
SDP
Catalog No. 50366182
Encompass_Preferred
Hydromount is the traditional choice whenever a nonfluorescing aqueous medium is needed. Hydromount is water-based and is suitable for mounting specimens that have been processed in water. Hydromount is effective for frozen sections, amyloid, and immunofluorescent staining procedures. Should it become necessary, Hydromount may be removed by soaking the slides in warm saline.
Catalog No. 50-366-182 Supplier Electron Microscopy Sciences Supplier No. 17966
Hydromount is the traditional choice whenever a nonfluorescing aqueous medium is needed. Hydromount is water-based and is suitable for mounting specimens that have been processed in water. Hydromount is effective for frozen sections, amyloid, and immunofluorescent staining procedures. Should it become necessary, Hydromount may be removed by soaking the slides in warm saline.