Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydronopol | 4747-61-9 | 98.09% | 168.28 | 100 G
SDP

Catalog No. 5000364914
Encompass_Preferred

Hydronopol is a saturated alcohol terpene derivative. It exhibits central inhibitory, anticonvulsant, analgesic, anticholinergic, and antihypertensive activities, making it suitable for research related to neurological and cardiovascular diseases.

  • Saturated alcohol terpene derivative
  • Central inhibitory activity
  • Anticonvulsant activity
  • Analgesic activity
  • Anticholinergic activity
  • Antihypertensive activity
  • Used in neurological disease research
  • Used in cardiovascular disease research
  • Purity 98.09%
  • Molecular Formula: C11H20O
  • Molecular Weight: 168.28
  • CAS Number: 4747-61-9

Catalog No. 50-003-64914 Supplier Medchemexpress LLC Supplier No. 100GHYZ16070
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydronopol is a saturated alcohol terpene derivative. It exhibits central inhibitory, anticonvulsant, analgesic, anticholinergic, and antihypertensive activities, making it suitable for research related to neurological and cardiovascular diseases.

  • Saturated alcohol terpene derivative
  • Central inhibitory activity
  • Anticonvulsant activity
  • Analgesic activity
  • Anticholinergic activity
  • Antihypertensive activity
  • Used in neurological disease research
  • Used in cardiovascular disease research
  • Purity 98.09%
  • Molecular Formula: C11H20O
  • Molecular Weight: 168.28
  • CAS Number: 4747-61-9

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.