Learn More
Medchemexpress LLC Hydronopol | 4747-61-9 | HY-Z16070 | 98.1% | 168.28 | 25 G
SDP
Catalog No. 5000366103
Encompass_Preferred
Hydronopol is a saturated alcohol terpene derivative that exhibits central inhibitory, anticonvulsant, analgesic, anticholinergic, and antihypertensive activities. It can be used for research related to neurological and cardiovascular diseases.
  • Saturated alcohol terpene derivative
  • Central inhibitory activity
  • Anticonvulsant activity
  • Analgesic activity
  • Anticholinergic activity
  • Antihypertensive activity
  • Suitable for neurological disease research
  • Suitable for cardiovascular disease research
Catalog No. 50-003-66103 Supplier Medchemexpress LLC Supplier No. 25GHYZ16070
Hydronopol is a saturated alcohol terpene derivative that exhibits central inhibitory, anticonvulsant, analgesic, anticholinergic, and antihypertensive activities. It can be used for research related to neurological and cardiovascular diseases.
  • Saturated alcohol terpene derivative
  • Central inhibitory activity
  • Anticonvulsant activity
  • Analgesic activity
  • Anticholinergic activity
  • Antihypertensive activity
  • Suitable for neurological disease research
  • Suitable for cardiovascular disease research