Learn More
Bachem Fmoc-AEEAc-OH
SDP
Catalog No. 501947107
Encompass_Preferred
Fmoc-AEEAc-OH, 166108-71-0, C21H23NO6, Mr 385.42, 5g. 8-(Fmoc-amino)-3,6-dioxa-octanoic acid, Fmoc-Adoa-OH, {2-[2-(Fmoc-amino)-ethoxy]-ethoxy}-acetic acid, Fmoc-8-amino-3,6-dioxa-octanoic acid. Hydrophilic heterobifunctional spacer useful to attach chromophores. Building block for the synthesis of spacers of defined length.
Catalog No. 50-194-7107 Supplier Bachem Supplier No. 4037914.5000
Add to Cart
Add to Cart
Fmoc-AEEAc-OH, 166108-71-0, C21H23NO6, Mr 385.42, 5g. 8-(Fmoc-amino)-3,6-dioxa-octanoic acid, Fmoc-Adoa-OH, {2-[2-(Fmoc-amino)-ethoxy]-ethoxy}-acetic acid, Fmoc-8-amino-3,6-dioxa-octanoic acid. Hydrophilic heterobifunctional spacer useful to attach chromophores. Building block for the synthesis of spacers of defined length.