Learn More
Electron Microscopy Sciences Hydrophobic PTFE Printed Microscope Slide 30 Round Wells, 2 mm
SDP
Catalog No. 5029202
Encompass_Preferred
"PTFE" Printed Slides, are extremely hydrophobic, acetone proof, chemical resistant, autoclavable, and precleaned. They are perfect for controlling cross-contamination in staining, immunolabeling and immunofluorescence tests. The well surface is 100% wettable, which enhances cell attachment and increases its spreading capabilities. These slides minimize the use of expensive purified antigens and antibodies.
The shelf life ranges from 18 months to two years. However, it is recommended that all glass slides (Printed or Un-printed) be used in a timely manner to avoid glass "weathering", (slides becoming cloudy or hazy even with proper storage. All of our Printed Slides are autoclavable and have a frosted end. Intended for one time use only. 30 round wells, hydrophobic, Measures: 3x1" (76x26x1mm).
Size: 2 mm
Catalog No. 50-292-02 Supplier Electron Microscopy Sciences Supplier No. 6343402
"PTFE" Printed Slides, are extremely hydrophobic, acetone proof, chemical resistant, autoclavable, and precleaned. They are perfect for controlling cross-contamination in staining, immunolabeling and immunofluorescence tests. The well surface is 100% wettable, which enhances cell attachment and increases its spreading capabilities. These slides minimize the use of expensive purified antigens and antibodies.
The shelf life ranges from 18 months to two years. However, it is recommended that all glass slides (Printed or Un-printed) be used in a timely manner to avoid glass "weathering", (slides becoming cloudy or hazy even with proper storage. All of our Printed Slides are autoclavable and have a frosted end. Intended for one time use only. 30 round wells, hydrophobic, Measures: 3x1" (76x26x1mm).
Size: 2 mm