Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Hydroponics 4-Way Test Kit
SDPSCI_ED

Catalog No. s43406 Shop All LaMotte Company Products
Change view
Click to view available options
Quantity:
Reagents sufficient for 50 tests per factor
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S43406 Reagents sufficient for 50 tests per factor
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S43406 Supplier LaMotte™ Supplier No. 356101
Only null left
Add to Cart
Add to Cart

Includes

Instructions and 75-page hydroponics handbook

Lamotte™ Hydroponics 4-Way Tesk Kit offers tests for pH and three key nutrient factors: nitrogen, phosphorus and potassium

Test kit allows the hydroponic hobbyist to maintain proper nutrient balance and to achieve optimum growing conditions in soil-less cultures.

Octa-Slide Comparator Tests

  • pH: pH 4.5 to 8.0
  • Nitrate Nitrogen: 5 to 200ppm (by dilution)
  • Phosphorus: 3 to 30ppm
  • Sulfate: 0 to 200ppm

Direct Reading Turbidity Tube

  • Potassium: 0 to 250ppm

Specifications

Includes Instructions and 75-page hydroponics handbook
Quantity Reagents sufficient for 50 tests per factor
Product Type Hydroponics Test Kit
Description Complete labware are foam-mounted in a sturdy carrying case
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.