Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

LaMotte™ Hydroponics Kit Reagent Refill
SDPSCI_ED

Catalog No. S43410 Shop All LaMotte Company Products
Change view
Click to view available options
Grade:
5 to 15
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S43410 5 to 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S43410 Supplier LaMotte™ Supplier No. R3561
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reagent refill package for the HP-1 Hydroponics kit.

  • Contains all reagents and consumables as supplied in the original kit.

Specifications

Grade 5 to 15
Age 12 to 20
For Use With (Equipment) HP-1 Hydroponics Kit
Weight (English) 1 lb.
Product Type Chemical Test Reagents
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.