Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Hydroquinidine | 1435-55-8 | 100.0% | 25 G
SDP

Catalog No. 5000406830
Encompass_Preferred

Hydroquinidine, also known as Dihydroquinidine, is a potent ion channel blocker with demonstrated anti-cancer activity against colon, pancreatic, and hepatocellular cancer cells. It is also known to prolong the QT interval and exhibit antiarrhythmic efficacy.

  • Potent ion channel blocker
  • Demonstrates strong anti-cancer activity on colon, pancreatic, and hepatocellular cancer cells
  • Prolongs the QT interval
  • Exhibits antiarrhythmic efficacy
  • Molecular formula: C20H26N2O2
  • Molecular weight: 326.44
  • Appearance: White to off-white solid
  • For research use only

Catalog No. 50-004-06830 Supplier Medchemexpress LLC Supplier No. 25GHYB0997
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Hydroquinidine, also known as Dihydroquinidine, is a potent ion channel blocker with demonstrated anti-cancer activity against colon, pancreatic, and hepatocellular cancer cells. It is also known to prolong the QT interval and exhibit antiarrhythmic efficacy.

  • Potent ion channel blocker
  • Demonstrates strong anti-cancer activity on colon, pancreatic, and hepatocellular cancer cells
  • Prolongs the QT interval
  • Exhibits antiarrhythmic efficacy
  • Molecular formula: C20H26N2O2
  • Molecular weight: 326.44
  • Appearance: White to off-white solid
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.