Learn More
Medchemexpress LLC Hydroquinidine | 1435-55-8 | 99.9% | 1 ML
SDP
Catalog No. 5000407557
Encompass_Preferred
Hydroquinidine (Dihydroquinidine) is a potent ion channel blocker that demonstrates strong anti-cancer activity against colon, pancreatic, and hepatocellular cancer cells. It also prolongs the QT interval and possesses antiarrhythmic efficacy.
  • Potent ion channel blocker
  • Exhibits strong anti-cancer activity on colon, pancreatic, and hepatocellular cancer cells
  • Prolongs the QT interval
  • Possesses antiarrhythmic efficacy
  • For research use only
Catalog No. 50-004-07557 Supplier Medchemexpress LLC Supplier No. HYB099710MM/1ML
Hydroquinidine (Dihydroquinidine) is a potent ion channel blocker that demonstrates strong anti-cancer activity against colon, pancreatic, and hepatocellular cancer cells. It also prolongs the QT interval and possesses antiarrhythmic efficacy.
  • Potent ion channel blocker
  • Exhibits strong anti-cancer activity on colon, pancreatic, and hepatocellular cancer cells
  • Prolongs the QT interval
  • Possesses antiarrhythmic efficacy
  • For research use only